Sodium tellurite(IV) ≥99.5% (metals basis)
Supplier: BeanTown Chemical
Synonyms:
Sodium tellurite
, Disodium trioxotellurate
225310-10G
225310-50G
BT225310-10GEA
42.59
USD
BT225310-10G
BT225310-50G
Sodium tellurite(IV) ≥99.5% (metals basis)
Sodium tellurite(IV)
- RTECS: WY2450000
- -100 Mesh Powder
Formula:
Na₂O₃Te MW: 221.58 g/mol |
MDL Number:
MFCD00036141 CAS Number: 10102-20-2 EINECS: 233-268-4 UN: 3284 ADR: 6.1,II |
Specification Test Results
Assay | 99.5% min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...