Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-α-bromoisobutyrate ≥99%

Supplier: BeanTown Chemical


225275-100ML
BT225275-100MLEA 36.8 USD InStock
BT225275-100ML
Methyl-α-bromoisobutyrate ≥99%
Methyl-α-bromoisobutyrate
CAS: 23426-63-3; EC No: 245-657-6; MDL No: MFCD00017854
UN No: UN2929; Haz Class: 6.1 (3); Packing Group: II
Liquid; Linear Formula: (CH3)2CBrCOOCH3; Molecular Formula: C5H9BrO2; MW: 181.03
Flash point: 49°C (120°F)
Density (g/mL): 1.399; Refractive Index: 1.451


Formula: C₅H₉BrO₂
MW: 181.029 g/mol
CAS Number: 23426-63-3
Order Now
Specification Test Results
Assay 99%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty