Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Indium(III) nitrate hydrate ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Indium trinitrate hydrate

225185-10G 225185-50G
BT225185-10GEA 59.65 USD InStock
BT225185-10G BT225185-50G
Indium(III) nitrate hydrate ≥99.99% (trace metals basis)
Indium (III) nitrate hydrate
CAS: 207398-97-8; EC No: 237-393-5; MDL No: MFCD00149742
UN No: UN1477; Haz Class: 5.1; Packing Group: III
Powder/Lump; Linear Formula: In(NO3)3·xH2O; MW: 300.83 (anhydrous)
Hygroscopic


Formula: InN₃O₉·H₂O
MW: 318,85 g/mol
MDL Number: MFCD00149742
CAS Number: 207398-97-8
EINECS: 237-393-5
UN: 1477
ADR: 5.1,III
Order Now
Specification Test Results
Assay 99.99%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty