Trifluoroacetamidine ≥85%, Technical Grade

Supplier: BeanTown Chemical

Synonyms: 2,2,2-Trifluoroacetamidine

225170-25G 225170-5G 225170-100G
BT225170-5GEA 83.75 USD
BT225170-5G BT225170-100G BT225170-25G
Trifluoroacetamidine ≥85%, Technical Grade
Trifluoroacetamidine

  • Refractive index: 1.382
  • Moisture Sensitive
  • Liquid

Formula: C₂H₃F₃N₂
Boiling Pt: 35…37 °C/11 mmHg
Density: 1.45 g/mL
MDL Number: MFCD00041545
CAS Number: 354-37-0
EINECS: 000-000-0
UN: 3267
ADR: 8,II

Order Now

Specification Test Results

Assay 85%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR