Molybdenum(VI) fluoride

Supplier: BeanTown Chemical

225130-50G
BT225130-50GEA 1782.42 USD
BT225130-50G
Molybdenum(VI) fluoride
Molybdenum(VI) fluoride

  • RTECS: QA4681450
  • Moisture Sensitive
  • Liquid

Formula: F₆Mo
Boiling Pt: 37 °C
Melting Pt: 17 °C
Density: 2.3 g/mL
MDL Number: MFCD00014220
CAS Number: 7783-77-9
EINECS: 232-026-5
ADR: 6.1 (8),II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR