2,3-Dimethylquinoxaline ≥97%

Supplier: BeanTown Chemical

225035-100G 225035-25G 225035-5G
BT225035-5GEA 41.4 USD
BT225035-5G BT225035-25G BT225035-100G
2,3-Dimethylquinoxaline ≥97%
2,3-Dimethylquinoxaline

  • RTECS: VD1960000
  • Solid

Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Melting Pt: 104…108 °C
MDL Number: MFCD00006728
CAS Number: 2379-55-7
EINECS: 219-162-0
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR