p-Ethyltoluene ≥97%

Supplier: BeanTown Chemical

Synonyms: 1-Ethyl-4-methylbenzene4-Methylethylbenzene4-Ethyltoluene

225020-100G 225020-25G 225020-5G
BT225020-25GEA 82.79 USD
BT225020-25G BT225020-100G BT225020-5G
p-Ethyltoluene ≥97%
p-Ethyltoluene

  • RTECS: XT2550000
  • Refractive index: 1.495
  • Liquid

Formula: C₉H₁₂
MW: 120.19 g/mol
Boiling Pt: 162 °C
Density: 0.861 g/mL
Flash Pt: 43 °C (109 °F)
MDL Number: MFCD00009263
CAS Number: 622-96-8
EINECS: 210-761-2
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR