Tungsten(VI) ethoxide 5% (w/v) in ethanol , 99.5% (trace metals basis)
Supplier: BeanTown Chemical
224925-100ML
224925-25ML
BT224925-100MLEA
411.33
USD
BT224925-100ML
BT224925-25ML
Tungsten(VI) ethoxide 5% (w/v) in ethanol , 99.5% (trace metals basis)
Tungsten (VI) ethoxide
- Moisture Sensitive
- Liquid
Formula:
C₁₀H₂₅O₅W MW: 454.21 g/mol |
MDL Number:
MFCD00156526 CAS Number: 62571-53-3 ADR: 3,II |
Specification Test Results
Assay | 99.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...