5-Chloropicolinonitrile ≥95%

Supplier: BeanTown Chemical

Synonyms: 5-Chloro-2-cyanopyridine

224795-10G
BT224795-10GEA 86.42 USD
BT224795-10G
5-Chloropicolinonitrile ≥95%
5-Chloropicolinonitrile

  • Solid

Formula: C₆H₃ClN₂
MW: 138.55 g/mol
Melting Pt: 106…108 °C
MDL Number: MFCD03788835
CAS Number: 89809-64-3
ADR: 6.1,III

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR