Aluminium ≥99.5% (trace metals basis), Powder -100+325 mesh

Supplier: BeanTown Chemical

Synonyms: Aluminum

224515-50G 224515-1KG 224515-250G
BT224515-50GEA 36.46 USD
BT224515-50G BT224515-1KG BT224515-250G
Aluminium ≥99.5% (trace metals basis), Powder -100+325 mesh
Aluminium

  • RTECS: BD0330000
  • Powder

Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2460 °C
Melting Pt: 660.4 °C
Density: 2.7 g/mL
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
UN: 1396
ADR: 4.3,II

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR