Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2-Dichlorobenzene ≥98.0% for spectrophotometry

Supplier: BeanTown Chemical

Synonyms: o-Dichlorobenzene

224500-4L 224500-1L 224500-4X1L
BT224500-4LEA 177.94 USD InStock
BT224500-4L BT224500-1L BT224500-4X1L
1,2-Dichlorobenzene ≥98.0% for spectrophotometry
1,2-Dichlorobenzene
CAS: 95-50-1; EC No: 202-425-9; MDL No: MFCD00000535; RTECS: CZ4500000
UN No: UN1591; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: C6H4Cl2; MW: 147.00
Melting Point: -18°; Boiling Point: 178-180°; Flash point: 66°C (151°F)
Density (g/mL): 1.306; Refractive Index: 1.551


Formula: C₆H₄Cl₂
MW: 147 g/mol
Boiling Pt: 180.5 °C (1013 hPa)
Melting Pt: -17 °C
Density: 1.309 g/cm³ (20 °C)
Flash Pt: 66 °C
Storage Temperature: ambient
MDL Number: MFCD00000535
CAS Number: 95-50-1
EINECS: 202-425-9
UN: 1591
ADR: 6.1,III
Merck Index: 13,03081
Order Now
Specification Test Results
Assay 98.0% min.


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty