Thiophenol ≥99%

Supplier: BeanTown Chemical

224440-50G 224440-250G 224440-10G
BT224440-10GEA 28.68 USD
BT224440-10G BT224440-50G BT224440-250G
Thiophenol ≥99%
Thiophenol

  • RTECS: DC0525000
  • Refractive index: 1.588
  • Air Sensitive
  • Liquid

Formula: C₆H₆S
MW: 110.18 g/mol
Boiling Pt: 169 °C
Melting Pt: –15 °C
Density: 1.073 g/mL
Flash Pt: 50 °C (122 °F)
MDL Number: MFCD00004826
CAS Number: 108-98-5
EINECS: 203-635-3
UN: 2337
ADR: 6.1,I

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR