Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thallium(I) acetate ≥99.995% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Thallous acetateAcetic acid thallium (I) salt

224255-100G 224255-25G
BT224255-100GEA 743.33 USD InStock
BT224255-100G BT224255-25G
Thallium(I) acetate ≥99.995% (trace metals basis)
Thallium (I) acetate
CAS: 563-68-8; EC No: 209-257-5; MDL No: MFCD00013045
UN No: UN1707; Haz Class: 6.1; Packing Group: II
Crystalline; Linear Formula: CH3COOTl; Molecular Formula: C2H3O2Tl; MW: 263.42
Melting Point: 126°
Density (g/mL): 3.68
Hygroscopic


Formula: C₂H₃O₂Tl
MW: 263,43 g/mol
Boiling Pt: 1457 °C (1013 hPa)
Melting Pt: 110 °C
Density: 3.68 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00013045
CAS Number: 563-68-8
EINECS: 209-257-5
UN: 1707
ADR: 6.1,II
Merck Index: 12,09392
Order Now
Specification Test Results
Assay 99.995%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty