N,N-Dimethylaminophosphoryl dichloride ≥97%
Supplier: BeanTown Chemical
Synonyms:
N,N-Dimethylphosphoramidodichloridate
224235-5G
224235-25G
BT224235-25GEA
160.96
USD
BT224235-25G
BT224235-5G
N,N-Dimethylaminophosphoryl dichloride ≥97%
N,N-Dimethylaminophosphoryl dichloride
- RTECS: TB4025000
- Refractive index: 1.464
- Air Sensitive, Hygroscopic, Moisture Sensitive
- Liquid
Formula:
C₂H₆Cl₂NOP Boiling Pt: 77…79 °C/11 mmHg Density: 1.362 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00013896 CAS Number: 677-43-0 EINECS: 211-641-2 UN: 3265 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...