Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Yttrium (III) nitrate hexahydrate ≥99.9% (trace rare earth metals basis)

Supplier: BeanTown Chemical

Synonyms: Yttrium trinitrate hexahydrate

224125-25G 224125-5G 224125-100G
BT224125-25GEA 38.42 USD InStock
BT224125-25G BT224125-5G BT224125-100G
Yttrium (III) nitrate hexahydrate ≥99.9% (trace rare earth metals basis)
Yttrium (III) nitrate hexahydrate
CAS: 13494-98-9; EC No: 233-802-6; MDL No: MFCD00149940
UN No: UN1477; Haz Class: 5.1; Packing Group: III
Crystalline; Linear Formula: Y(NO3)3·6H2O; MW: 383.01
Density (g/mL): 2.682
Hygroscopic


Formula: N₃O₉Y·6H₂O
MW: 383,01 g/mol
Melting Pt: 51.8 °C
Density: 2.682 g/cm³ (20 °C)
MDL Number: MFCD00149940
CAS Number: 13494-98-9
EINECS: 233-802-6
UN: 1477
ADR: 5.1,III
Order Now
Specification Test Results
Assay 99.9%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty