Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lead selenide ≥99.999% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Lead(II) selenide

224055-100G 224055-25G
BT224055-100GEA 745.58 USD InStock
BT224055-100G BT224055-25G
Lead selenide ≥99.999% (trace metals basis)
Lead selenide
CAS: 12069-00-0; EC No: 235-109-4; MDL No: MFCD00016273
UN No: UN3077; Haz Class: 9; Packing Group: III
3-6mm Fused Lump; Molecular Formula: PbSe ; MW: 286.15
Density (g/mL): 8.1


Formula: PbSe
MW: 286,16 g/mol
Melting Pt: 79.5 to 80 °C
Density: 8.1 g/cm³ (20 °C)
MDL Number: MFCD00016273
CAS Number: 12069-00-0
EINECS: 235-109-4
UN: 3283
ADR: 6.1,III
Order Now
Specification Test Results
Assay 99.999%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty