BEAN224045

224045-2.5L
BT224045-2.5LEA 326.32 USD
BT224045-2.5L
BEAN224045
  • RTECS: GW0220000
  • Refractive index: 1.462
  • Liquid
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR