Hexachloroiridic(IV) acid hydrate ≥99% (trace metals basis), lumps

Supplier: BeanTown Chemical

Synonyms: Dihydrogen hexachloroiridate(IV) hydrateHexachloroiridium(IV) acid hydrateHydrogen hexachloroiridate(IV) hydrateHydrogen iridium hexachloride

223680-25G 223680-5G 223680-1G
BT223680-1GEA 265.29 USD
BT223680-1G BT223680-5G BT223680-25G
Hexachloroiridic(IV) acid hydrate ≥99% (trace metals basis), lumps
Hexachloroiridic (IV) acid
Formula: H₂Cl₆Ir·xH₂O
MW: 406.93 g/mol (anhy)
MDL Number: MFCD00149906
EINECS: 241-012-8
ADR: 8,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR