Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Titanium(IV) butanolate ≥99%

Supplier: BeanTown Chemical

Synonyms: Titanium tetrabutanolateTitanium (IV) butoxideTetra-n-butyl orthotitanateTitanium (IV) n-butoxide

223610-500G 223610-100G
BT223610-500GEA 67.57 USD InStock
BT223610-500G BT223610-100G
Titanium(IV) butanolate ≥99%
Titanium(IV) butanolate
CAS: 5593-70-4; EC No: 227-006-8; MDL No: MFCD00009433; RTECS: XR1585000
UN No: UN2924; Haz Class: 3(8); Packing Group: III
Liquid; Linear Formula: Ti[O(CH2)3CH3]4; Molecular Formula: C16H36O4Ti; MW: 340.36
Melting Point: -55°; Boiling Point: 310-314°; Flash point: 41°C (106°F)
Density (g/mL): 0.998; Refractive Index: 1.491
Moisture Sensitive


Formula: C₁₆H₃₆O₄Ti
MW: 340,34 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: -55 °C
Density: 0.993 g/cm³ (20 °C)
Flash Pt: 47 °C
MDL Number: MFCD00009433
CAS Number: 5593-70-4
EINECS: 227-006-8
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Assay 99%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty