Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc Dimethyldithiocarbamate ≥97%

Supplier: BeanTown Chemical

Synonyms: Dimethyldithiocarbamic Acid Zinc SaltZiram

223595-1KG
BT223595-1KGEA 92.8 USD InStock
BT223595-1KG
Zinc Dimethyldithiocarbamate ≥97%
Zinc dimethyldithiocarbamate
CAS: 137-30-4; EC No: 205-288-3; MDL No: MFCD00064797; RTECS: ZH0525000
UN No: UN2811; Haz Class: 6.1; Packing Group: II
Powder; Linear Formula: [(CH3)2NCSS]2Zn; Molecular Formula: C6H12N2S4Zn; MW: 305.82
Melting Point: 248-257°


Formula: C₃H₇NS₂
MW: 305,80 g/mol
MDL Number: MFCD00064797
CAS Number: 137-30-4
Order Now
Specification Test Results
Assay 97%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty