1,3-Di-o-tolylguanidine ≥99%

Supplier: BeanTown Chemical

223515-500G 223515-100G 223515-25G
BT223515-100GEA 56.71 USD
BT223515-100G BT223515-500G BT223515-25G
1,3-Di-o-tolylguanidine ≥99%
1,3-Di-o-tolylguanidine

  • RTECS: MF1400000
  • Solid

Formula: C₁₅H₁₇N₃
MW: 239.32 g/mol
Melting Pt: 176…178 °C
MDL Number: MFCD00008513
CAS Number: 97-39-2
EINECS: 202-577-6
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR