Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium acetylide

Supplier: BeanTown Chemical

Synonyms: Calcium acetylideCalcium carbide

223480-1KG
BT223480-1KGEA 64.27 USD InStock
BT223480-1KG
Calcium acetylide
Calcium acetylide
CAS: 75-20-7; EC No: 200-848-3; MDL No: MFCD00010905
UN No: UN1402; Haz Class: 4.3; Packing Group: II
9-40mm Lump; Linear Formula: CaC2; MW: 64.10
Density (g/mL): 2.22
Moisture Sensitive


Formula: CaC₂
MW: 64,1 g/mol
Melting Pt: 2300 °C
Density: 2.22 g/cm³ (20 °C)
MDL Number: MFCD00010905
CAS Number: 75-20-7
EINECS: 200-848-3
UN: 1402
ADR: 4.3,II
Merck Index: 12,01696
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty