Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mercury(I) nitrate dihydrate ≥98%

Supplier: BeanTown Chemical

Synonyms: Dimercury dinitrate dihydrateMercurous nitrate dihydrate

223415-1KG 223415-250G 223415-50G
BT223415-1KGEA 567.62 USD InStock
BT223415-1KG BT223415-250G BT223415-50G
Mercury(I) nitrate dihydrate ≥98%
Mercury (I) nitrate dihydrate
CAS: 14836-60-3; EC No: 233-886-4; MDL No: MFCD00150812; RTECS: OW8000000
UN No: UN1627; Haz Class: 6.1; Packing Group: II
Crystalline; Linear Formula: Hg2(NO3)2·2H2O; MW: 561.22
Melting Point: 70°
Density (g/mL): 4.78
Light Sensitive


Formula: Hg₂N₂O₆·2H₂O
MW: 561,22 g/mol
Melting Pt: 70 °C
Density: 4.78 g/cm³ (20 °C)
MDL Number: MFCD00150812
CAS Number: 14836-60-3
EINECS: 233-886-4
UN: 1627
ADR: 6.1,II
Merck Index: 13,05923
Order Now
Specification Test Results
Assay 98%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty