Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Aminophenyl phenyl sulfide ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Aminodiphenyl Sulfide2-(Phenylthio)aniline2-Aminodiphenyl Sulphide

223390-100G
BT223390-100GEA 87.63 USD InStock
BT223390-100G
2-Aminophenyl phenyl sulfide ≥98%
2-Aminophenyl phenyl sulfide
CAS: 1134-94-7; EC No: 413-030-8; MDL No: MFCD00190139
UN No: UN3077; Haz Class: 9; Packing Group: III
Crystalline; Molecular Formula: C12H11NS; MW: 201.29
Melting Point: 43°; Flash point: 175°C (347°F)
Air Sensitive


Formula: C₁₂H₁₁NS
MW: 201,29 g/mol
CAS Number: 1134-94-7
Order Now
Specification Test Results
Assay 98%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty