Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dicyclohexylchlorophosphine ≥98%

Supplier: BeanTown Chemical

Synonyms: Chlorodicyclohexylphosphine

223370-1G 223370-5G
BT223370-1GEA 65.55 USD InStock
BT223370-1G BT223370-5G
Dicyclohexylchlorophosphine ≥98%
Dicyclohexylchlorophosphine
CAS: 16523-54-9; MDL No: MFCD00014294
UN No: UN3265; Haz Class: 8; Packing Group: II
Liquid; Molecular Formula: C12H22ClP ; MW: 232.73
Boiling Point: 165°/12 mmHg; Flash point: 110°C (230°F)
Density (g/mL): 1.054; Refractive Index: 1.533
Air Sensitive, Moisture Sensitive


Formula: C₁₂H₂₂ClP
MW: 232,73 g/mol
MDL Number: MFCD00014294
CAS Number: 16523-54-9
EINECS: 000-000-0
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Assay 98%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty