Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tetra-Chloroauric(III) acid (40 - 44% Au) (w/w) in aqueous solution

Supplier: BeanTown Chemical

Synonyms: Gold Chloridetetra-Chloroauric acidHydrogen tetrachloroaurate (III)

223355-C1G 223355-C5G
BT223355-C1GEA 206.1 USD InStock
BT223355-C1G BT223355-C5G
tetra-Chloroauric(III) acid (40 - 44% Au) (w/w) in aqueous solution
tetra-Chloroauric (III) acid
CAS: 16903-35-8; EC No: 240-948-4; MDL No: MFCD00011322; RTECS: CK6330000
UN No: UN3264; Haz Class: 8; Packing Group: III
Liquid; Molecular Formula: HAuCl4; MW: 339.79
Light Sensitive


Formula: HAuCl₄
MW: 339,78 g/mol
Density: 3.9 g/cm³ (25 °C)
MDL Number: MFCD00011322
CAS Number: 16903-35-8
EINECS: 240-948-4
UN: 3264
ADR: 8,III
Merck Index: 13,04536
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty