Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hexane (mixture of isomers) ≥99.0% for HPLC

Supplier: BeanTown Chemical

Danger
Synonyms: HexanesIsohexane

223335-4X1L 223335-4L 223335-1L
BT223335-4X1LEA 131.78 USD InStock
BT223335-4X1L BT223335-4L BT223335-1L
Hexane (mixture of isomers) ≥99.0% for HPLC
Hexane (mixture of isomers)
CAS: 92112-69-1; EC No: 295-570-2; MDL No: MFCD02179311
UN No: UN1208; Haz Class: 3; Packing Group: II
Liquid; Molecular Formula: C6H14; MW: 86.18
Melting Point: -95°; Boiling Point: 68-70°; Flash point: -23°C (-9°F)
Density (g/mL): 0.672; Refractive Index: 1.379


Formula: H₃C(CH₂)₄CH₃
MW: 86,18 g/mol
Boiling Pt: 68 to 70 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.66 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://us.vwr.com/stibo/web/std.lang.all/05/68/16640568.jpg
Order Now
Specification Test Results
Assay 99.0% min.


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty