Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

p-Toluenesulfonic acid monohydrate ≥98%

Supplier: BeanTown Chemical

Synonyms: Toluene-4-sulphonic acid monohydrateToluene-4-sulfonic acid monohydrate

223285-5KG 223285-500G 223285-100G
BT223285-5KGEA 191.47 USD InStock
BT223285-5KG BT223285-500G BT223285-100G
p-Toluenesulfonic acid monohydrate ≥98%
p-Toluenesulfonic acid monohydrate
CAS: 6192-52-5; EC No: 203-180-0; MDL No: MFCD00142137; RTECS: XT6300000
UN No: UN2585; Haz Class: 8; Packing Group: III
Solid; Linear Formula: CH3C6H4SO3H·H2O; Molecular Formula: C7H8O3S·H2O; MW: 190.22
Melting Point: 103-106°; Boiling Point: 140°/22 mmHg; Flash point: 150°C (302°F)
Density (g/mL): 1.24
Hygroscopic


Formula: C₇H₈O₃S·1H₂O
MW: 190,22 g/mol
Melting Pt: 102 to 104 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 180 °C
Storage Temperature: ambient
MDL Number: MFCD00142137
CAS Number: 6192-52-5
EINECS: 203-180-0
UN: 2585
ADR: 8,III
Merck Index: 12,09671
Order Now
Specification Test Results
Assay 98%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty