Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lead(II) iodide ≥99.995% (trace metals basis)

Supplier: BeanTown Chemical


223265-25G 223265-100G
BT223265-25GEA 70.25 USD InStock
BT223265-25G BT223265-100G
Lead(II) iodide ≥99.995% (trace metals basis)
Lead (II) iodide
CAS: 10101-63-0; EC No: 233-256-9; MDL No: MFCD00011163
UN No: UN2291; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: PbI2; MW: 461.00
Melting Point: 402°; Boiling Point: 954°
Density (g/mL): 6.16
Light Sensitive


Formula: I₂Pb
MW: 461,01 g/mol
Boiling Pt: 954 °C (1013 hPa)
Melting Pt: 402 °C
Density: 6.16 g/cm³ (20 °C)
MDL Number: MFCD00011163
CAS Number: 10101-63-0
EINECS: 233-256-9
UN: 2291
ADR: 6.1,III
Merck Index: 12,05430
Order Now
Specification Test Results
Assay 99.995%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty