1,2,3,4-Tetrahydronaphthalene ≥97%
Supplier: BeanTown Chemical
223235-250G
223235-5KG
223235-1KG
BT223235-5KGEA
200.55
USD
BT223235-5KG
BT223235-1KG
BT223235-250G
1,2,3,4-Tetrahydronaphthalene ≥97%
1,2,3,4-Tetrahydronaphthalene
- RTECS: QK3850000
- Refractive index: 1.541
- Air Sensitive
- Liquid
Formula:
C₁₀H₁₂ MW: 132.21 g/mol Boiling Pt: 207 °C Melting Pt: –35 °C Density: 0.973 g/mL Flash Pt: 71 °C (160 °F) |
MDL Number:
MFCD00001733 CAS Number: 119-64-2 EINECS: 204-340-2 UN: 3082 ADR: 9,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...