Hydrazinium monochloride ≥97%
Supplier: BeanTown Chemical
Synonyms:
Hydrazine monohydrochloride
, Hydrazinium chloride
, Hydrazine hydrochloride
, Hydrazinium monochloride
223110-250G
BT223110-250GEA
69.8
USD
BT223110-250G
Hydrazinium monochloride ≥97%
Hydrazinium monochloride
- RTECS: MV4625000
- Solid
Formula:
H₅ClN₂ MW: 68.51 g/mol Boiling Pt: 240 °C Melting Pt: 89 °C |
MDL Number:
MFCD00044368 CAS Number: 2644-70-4 EINECS: 220-154-4 UN: 2923 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...