Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rubidium nitrate ≥99% (trace metals basis)

Supplier: BeanTown Chemical


223065-25G 223065-100G
BT223065-25GEA 69.6 USD InStock
BT223065-25G BT223065-100G
Rubidium nitrate ≥99% (trace metals basis)
Rubidium nitrate
CAS: 13126-12-0; EC No: 236-060-1; MDL No: MFCD00011193
UN No: UN1477; Haz Class: 5.1; Packing Group: III
Powder; Linear Formula: RbNO3; MW: 147.47
Melting Point: 310°
Density (g/mL): 3.11; Refractive Index: 1.52
Hygroscopic


Formula: NO₃Rb
MW: 147,47 g/mol
Boiling Pt: 578 °C (1013 hPa)
Melting Pt: 310 °C
Density: 3.11 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00011193
CAS Number: 13126-12-0
EINECS: 236-060-1
UN: 1477
ADR: 5.1,III
Order Now
Specification Test Results
Assay 99%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty