Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Ethylimidazole ≥95%

Supplier: BeanTown Chemical

Synonyms: N-Ethyl-1H-imidazoleN-Ethylimidazole1-Ethyl-1H-imidazole

223045-100G 223045-500G
BT223045-100GEA 46.88 USD InStock
BT223045-100G BT223045-500G
1-Ethylimidazole ≥95%
1-Ethylimidazole
CAS: 7098-07-9; EC No: 230-403-9; MDL No: MFCD00055389
UN No: UN3267; Haz Class: 8; Packing Group: III
Liquid; Molecular Formula: C5H8N2; MW: 96.13
Melting Point: -27°; Boiling Point: 104°/ 26 mmHg; Flash point: 91°C (196°F)
Density (g/mL): 0.995


Formula: C₅H₈N₂
MW: 96.13 g/mol
Density: 0.995 g/cm³ (20 °C)
Storage Temperature: ambient
CAS Number: 7098-07-9
EINECS: 230-403-9
Order Now
Specification Test Results
Assay 95%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty