Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium cerium(IV) nitrate ≥99.5% (trace rare earth metals basis)

Supplier: BeanTown Chemical

Synonyms: Ammonium hexanitratocerate (IV)Ammonium ceric nitrateCerium (IV) ammonium nitrateCeric Ammonium NitrateDiammonium hexanitratocerate

222830-500G 222830-100G 222830-25G
BT222830-500GEA 248.92 USD InStock
BT222830-500G BT222830-100G BT222830-25G
Ammonium cerium(IV) nitrate ≥99.5% (trace rare earth metals basis)
Ammonium cerium (IV) nitrate
CAS: 16774-21-3; EC No: 240-827-6; MDL No: MFCD00151121
UN No: UN1477; Haz Class: 5.1; Packing Group: II
Crystalline; Linear Formula: (NH4)2Ce(NO3)6; MW: 548.23


Formula: (NH₄)₂Ce(NO₃)₆
MW: 548,22 g/mol
Melting Pt: 107 to 108 °C
Density: 2.2 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00151121
CAS Number: 16774-21-3
EINECS: 240-827-6
UN: 1477
ADR: 5.1,III
Merck Index: 13,00509
Order Now
Specification Test Results
Assay 99.5%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty