Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hafnium(IV) chloride ≥98% (trace metals basis excluding Zr)

Supplier: BeanTown Chemical

Synonyms: Hafnium tetrachloride

222785-100G 222785-25G 222785-5G
BT222785-100GEA 172.31 USD InStock
BT222785-100G BT222785-25G BT222785-5G
Hafnium(IV) chloride ≥98% (trace metals basis excluding Zr)
Hafnium (IV) chloride
CAS: 13499-05-3; EC No: 236-826-5; MDL No: MFCD00011034
UN No: UN3260; Haz Class: 8; Packing Group: II
Powder; Linear Formula: HfCl4; MW: 320.30
Boiling Point: 320° (sublimes)
Moisture Sensitive


Formula: Cl₄Hf
MW: 320,3 g/mol
Melting Pt: 431.85 °C
MDL Number: MFCD00011034
CAS Number: 13499-05-3
EINECS: 236-826-5
UN: 3260
ADR: 8,II
Order Now
Specification Test Results
Assay 98%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty