Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Allyl bromide ≥99% stabilized

Supplier: BeanTown Chemical

Synonyms: 3-Propenyl bromide3-Bromopropene

222765-250G 222765-1KG
BT222765-250GEA 27.05 USD InStock
BT222765-250G BT222765-1KG
Allyl bromide ≥99% stabilized
Allyl bromide
CAS: 106-95-6; EC No: 203-446-6; MDL No: MFCD00000244; RTECS: UC7090000
UN No: UN1099; Haz Class: 3 (6.1); Packing Group: I
Liquid; Linear Formula: CH2=CHCH2Br; Molecular Formula: C3H5Br; MW: 120.98
Melting Point: -119°; Boiling Point: 70-71°; Flash point: -1°C (30°F)
Density (g/mL): 1.398; Refractive Index: 1.469
Light Sensitive


Formula: C₃H₅Br
MW: 120,98 g/mol
Boiling Pt: 70 °C (1013 hPa)
Melting Pt: -119 °C
Density: 1.4 g/cm³ (20 °C)
Flash Pt: -1 °C
Storage Temperature: 2 - 8°C
MDL Number: MFCD00000244
CAS Number: 106-95-6
EINECS: 203-446-6
UN: 1099
ADR: 3,I
Merck Index: 13,00285
Order Now
Specification Test Results
Assay 99%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty