Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium phosphinate

Supplier: BeanTown Chemical


222750-500G 222750-100G
BT222750-500GEA 175.6 USD InStock
BT222750-500G BT222750-100G
Calcium phosphinate
Calcium phosphinate
CAS: 7789-79-9; EC No: 232-190-8; MDL No: MFCD00867083
UN No: UN3178; Haz Class: 4.1; Packing Group: II
Powder; Linear Formula: Ca(H2PO2)2; MW: 170.06
Hygroscopic


Formula: H₄CaO₄P₂
MW: 170,05 g/mol
MDL Number: MFCD00867083
CAS Number: 7789-79-9
EINECS: 232-190-8
UN: 3178
ADR: 4.1,II
Merck Index: 12,01718
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty