Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrahydrofuran ≥99.7% unstabilised for spectrophotometry

Supplier: BeanTown Chemical

Danger
Synonyms: OxolaneTHFCyclotetramethylene oxideTetramethylene oxide1,4-epoxy-ButaneOxacyclopentaneButane alpha,delta-oxide

222730-1L 222730-100ML 222730-4L
BT222730-1LEA 64.11 USD InStock
BT222730-1L BT222730-100ML BT222730-4L
Tetrahydrofuran ≥99.7% unstabilised for spectrophotometry
Tetrahydrofuran
CAS: 109-99-9; EC No: 203-726-8; MDL No: MFCD00005356; RTECS: LU5950000
UN No: UN2056; Haz Class: 3; Packing Group: II
Liquid, distilled in glass; Molecular Formula: C4H8O ; MW: 72.11
Melting Point: -108°; Boiling Point: 65-67°; Flash point: -17°C (1°F)
Density (g/mL): 0.889; Refractive Index: 1.407


Formula: C₄H₈O
MW: 72,11 g/mol
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: -108.5 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: -21.5 °C
Storage Temperature: ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://us.vwr.com/stibo/web/std.lang.all/95/19/789519.jpg
Order Now
Specification Test Results
Assay 99.7% min.


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty