Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gallium triiodide, anhydrous ≥99.999% (trace metals basis)

Supplier: BeanTown Chemical


Ampuled under argon

222665-25G 222665-5G 222665-1G
BT222665-25GEA 303.04 USD InStock
BT222665-25G BT222665-5G BT222665-1G
Gallium triiodide, anhydrous ≥99.999% (trace metals basis)
Gallium triiodide
CAS: 13450-91-4; EC No: 236-611-6; MDL No: MFCD00016107
UN No: UN3260; Haz Class: 8; Packing Group: II
Powder; Linear Formula: GaI3; MW: 450.43
Boiling Point: 345° (sublimes)
Density (g/mL): 4.15
Light Sensitive, Moisture Sensitive


Formula: GaI₃
MW: 450,44 g/mol
Boiling Pt: 345 °C (1013 hPa)
Melting Pt: 212 °C
Density: 4.15 g/cm³ (25 °C)
MDL Number: MFCD00016107
CAS Number: 13450-91-4
EINECS: 236-611-6
UN: 3260
ADR: 8,II
Order Now
Specification Test Results
Assay 99.999%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty