Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium fluoride ≥99%

Supplier: BeanTown Chemical


222620-1KG 222620-250G
BT222620-1KGEA 165.56 USD InStock
BT222620-1KG BT222620-250G
Barium fluoride ≥99%
Barium fluoride
CAS: 7787-32-8; EC No: 232-108-0; MDL No: MFCD00003450
UN No: UN1564; Haz Class: 6.1; Packing Group: III
-40 Mesh Powder ; Molecular Formula: BaF2; MW: 175.32
Boiling Point: 2260°
Density (g/mL): 4.89


Formula: BaF₂
MW: 175,33 g/mol
Boiling Pt: 2260 °C (1013 hPa)
Melting Pt: 1270 °C
Density: 4.89 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00003450
CAS Number: 7787-32-8
EINECS: 232-108-0
UN: 1564
ADR: 6.1,III
Merck Index: 12,01003
Order Now
Specification Test Results
Assay 99%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty