Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Manganese(II) chloride, anhydrous ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Manganous chlorideManganese dichloride

Packaged under argon.

222480-25G 222480-5G 222480-100G
BT222480-25GEA 200.48 USD InStock
BT222480-25G BT222480-5G BT222480-100G
Manganese(II) chloride, anhydrous ≥99.99% (trace metals basis)
Manganese (II) chloride
CAS: 7773-01-5; EC No: 231-869-6; MDL No: MFCD00011114; RTECS: OO9625000
UN No: UN3288; Haz Class: 6.1; Packing Group: III
-10 Mesh Beads; Linear Formula: MnCl2; MW: 125.84
Melting Point: 652°; Boiling Point: 1190°
Density (g/mL): 2.977
Hygroscopic


Formula: Cl₂Mn
MW: 125,84 g/mol
Boiling Pt: 1190 °C (1013 hPa)
Melting Pt: 650 °C
Density: 2.98 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00011114
CAS Number: 7773-01-5
EINECS: 231-869-6
UN: 3288
ADR: 6.1,III
Merck Index: 12,05768
Order Now
Specification Test Results
Assay 99.99%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty