Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tantalum pentachloride ≥99.8% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Tantalum (V) chloride

Packaged under argon.

222465-25G 222465-100G 222465-5G
BT222465-25GEA 102.6 USD InStock
BT222465-25G BT222465-100G BT222465-5G
Tantalum pentachloride ≥99.8% (trace metals basis)
Tantalum pentachloride
CAS: 7721-01-9; EC No: 231-755-6; MDL No: MFCD00011253; RTECS: WW5600000
UN No: UN3260; Haz Class: 8; Packing Group: III
Powder; Linear Formula: TaCl5; MW: 358.20
Melting Point: 216°; Boiling Point: 242°
Density (g/mL): 3.68
Moisture Sensitive


Formula: Cl₅Ta
MW: 358,21 g/mol
Boiling Pt: 232 to 235 °C (1013 hPa)
Melting Pt: 216 to 217 °C
Density: 3.68 g/cm³ (20 °C)
MDL Number: MFCD00011253
CAS Number: 7721-01-9
EINECS: 231-755-6
UN: 3260
ADR: 8,III
Order Now
Specification Test Results
Assay 99.8%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty