Ethanesulfonic acid ≥96%

Supplier: BeanTown Chemical

222420-250G 222420-50G 222420-10G
BT222420-250GEA 503.78 USD
BT222420-250G BT222420-50G BT222420-10G
Ethanesulfonic acid ≥96%
Ethanesulphonic acid

  • Refractive index: 1.435
  • Liquid

Formula: C₂H₆O₃S
MW: 110.13 g/mol
Boiling Pt: 123 °C/0.01 mmHg
Melting Pt: –17 °C
Density: 1.35 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00007529
CAS Number: 594-45-6
EINECS: 209-843-0
UN: 2586
ADR: 8,III

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR