Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium perrhenate ≥99% (trace metals basis)

Supplier: BeanTown Chemical


222325-500MG 222325-10G 222325-2G
BT222325-500MGEA 22.08 USD InStock
BT222325-500MG BT222325-10G BT222325-2G
Potassium perrhenate ≥99% (trace metals basis)
Potassium perrhenate
CAS: 10466-65-6; EC No: 233-953-8; MDL No: MFCD00011365; RTECS: SE1400000
UN No: UN1479; Haz Class: 5.1; Packing Group: II
Crystalline/Powder; Linear Formula: KReO4; MW: 289.30
Melting Point: 550°; Boiling Point: 1360-1370°
Density (g/mL): 4.887; Refractive Index: 1.643


Formula: KO₄Re
MW: 289,3 g/mol
Boiling Pt: 1370 °C (1013 hPa)
Melting Pt: 550 °C
Density: 4.88 g/cm³ (20 °C)
MDL Number: MFCD00011365
CAS Number: 10466-65-6
EINECS: 233-953-8
UN: 1479
ADR: 5.1,II
Order Now
Specification Test Results
Assay 99%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty