Dichloromethane ≥99.7% stabilized for HPLC

Synonyms: Methane dichlorideMethylene chloride

222305-1L 222305-4L 222305-250ML
BT222305-1LEA 50.12 USD
BT222305-1L BT222305-4L BT222305-250ML
Dichloromethane ≥99.7% stabilized for HPLC
Dichloromethane

  • RTECS: PA8050000
  • Refractive index: 1.424
  • Liquid

Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39…40 °C
Melting Pt: –97 °C
Density: 1.325 g/mL
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/14/30/20961430.jpg

Order Now

Specification Test Results

Assay 99.7% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR