2-Chloroethyl methacrylate ≥97% stabilized

222295-10G 222295-50G
BT222295-10GEA 90.89 USD
BT222295-10G BT222295-50G
2-Chloroethyl methacrylate ≥97% stabilized
2-Chloroethyl methacrylate

  • RTECS: OZ3855000
  • Liquid

Formula: C₆H₉ClO₂
Flash Pt: 69 °C (156 °F)
MDL Number: MFCD00045291
CAS Number: 1888-94-4
EINECS: 217-566-1
ADR: 6.1,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR