Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloropropanesulfonyl chloride ≥98%

Supplier: BeanTown Chemical


222285-250G 222285-50G 222285-10G
BT222285-250GEA 1072.2 USD InStock
BT222285-250G BT222285-50G BT222285-10G
3-Chloropropanesulfonyl chloride ≥98%
3-Chloropropanesulfonyl chloride
CAS: 1633-82-5; EC No: 216-646-3; MDL No: MFCD00007463; RTECS: TZ6670000
UN No: UN3265; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: Cl(CH2)3SO2Cl; Molecular Formula: C3H6Cl2O2S; MW: 177.05
Boiling Point: 70°/0.5 mmHg; Flash point: 110°C (230°F)
Density (g/mL): 1.456; Refractive Index: 1.489
Moisture Sensitive


Formula: C₃H₆Cl₂O₂S
MW: 177,039 g/mol
MDL Number: MFCD00007463
CAS Number: 1633-82-5
Order Now
Specification Test Results
Assay 98%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty