Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluoroethanol ≥95%

Supplier: BeanTown Chemical


222275-25G 222275-5G 222275-1G
BT222275-25GEA 183.57 USD InStock
BT222275-25G BT222275-5G BT222275-1G
2-Fluoroethanol ≥95%
2-Fluoroethanol
CAS: 371-62-0; EC No: 206-740-2; MDL No: MFCD00002828; RTECS: KL1575000
UN No: UN3384; Haz Class: 6.1 (3); Packing Group: I
Liquid; Linear Formula: FCH2CH2OH; Molecular Formula: C2H5FO; MW: 64.06
Melting Point: -27°; Boiling Point: 103°/757 mmHg; Flash point: 34°C (93°F)
Density (g/mL): 1.091; Refractive Index: 1.365
Air Sensitive


Formula: C₂H₅FO
MW: 64.06 g/mol
Boiling Pt: 100 to 102 °C (1013 hPa)
Melting Pt: -26.45 °C
Density: 1.002 g/cm³ (20 °C)
MDL Number: MFCD00002828
CAS Number: 371-62-0
EINECS: 206-740-2
UN: 3384
ADR: 6.1,I
Order Now
Specification Test Results
Assay 95%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty