Terbium ≥99.9% (trace rare earth metals basis), pieces
Supplier: BeanTown Chemical
222165-5G
222165-1G
BT222165-5GEA
296.93
USD
BT222165-5G
BT222165-1G
Terbium ≥99.9% (trace rare earth metals basis), pieces
Terbium
- Solid
Formula:
Tb MW: 158.93 g/mol Boiling Pt: 3230 °C Melting Pt: 1356 °C Density: 8.234 g/mL |
MDL Number:
MFCD00011256 CAS Number: 7440-27-9 EINECS: 231-137-6 ADR: 4.1,II |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...