Tetrachlorosilane ≥99% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Silicon tetrachlorideSilicon (IV) chloride

222150-10ML 222150-100ML 222150-1L
BT222150-1LEA 112.18 USD
BT222150-1L BT222150-100ML BT222150-10ML
Tetrachlorosilane ≥99% (trace metals basis)
Tetrachlorosilane

  • RTECS: VW0525000
  • Moisture Sensitive
  • Liquid

Formula: Cl₄Si
MW: 169.9 g/mol
Boiling Pt: 57.6 °C
Melting Pt: –70 °C
Density: 1.483 g/mL
MDL Number: MFCD00011229
CAS Number: 10026-04-7
EINECS: 233-054-0
UN: 1818
ADR: 8,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR